Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339429.1 | NP_001325635.1 | 11 - 12 | 2908 | 123 | |
NM_001339430.1 | NP_001325634.1 | 12 - 13 | 2859 | 123 | ||
NM_114820.4 | NP_566922.1 | 11 - 12 | 3019 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |