Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203116.2 | NP_001190045.1 | 1 - 2 | 52 | 98 | |
NM_001339445.1 | NP_001319718.1 | 1 - 2 | 188 | 98 | ||
NM_001339446.1 | NP_001325884.1 | 1 - 2 | 52 | 98 | ||
NM_114852.1 | NP_190561.1 | 1 - 2 | 52 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |