Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114860.4 | NP_190569.2 | 7 - 8 | 1059 | 119 | |
GenBank mRNA | BT000888.1 | - | 7 - 8 | 1022 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |