Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114860.4 | NP_190569.2 | 9 - 10 | 1191 | 77 | |
GenBank mRNA | BT000888.1 | - | 9 - 10 | 1154 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |