Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KICP-02 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339461.1 | NP_001326509.1 | 19 - 20 | 2412 | 116 | |
NM_001339462.1 | NP_001326510.1 | 18 - 19 | 2454 | 116 | ||
NM_001339463.1 | NP_001326508.1 | 19 - 20 | 2397 | 116 | ||
NM_114884.5 | NP_566931.1 | 19 - 20 | 2412 | 116 | ||
GenBank mRNA | AB061676.1 | - | 19 - 20 | 2387 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |