Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KICP-02 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339461.1 | NP_001326509.1 | 9 - 10 | 1392 | 124 | |
NM_001339462.1 | NP_001326510.1 | 8 - 9 | 1434 | 124 | ||
NM_001339463.1 | NP_001326508.1 | 9 - 10 | 1392 | 124 | ||
NM_114884.5 | NP_566931.1 | 9 - 10 | 1392 | 124 | ||
GenBank mRNA | AB061676.1 | - | 9 - 10 | 1367 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |