Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114896.5 | NP_190605.1 | 5 - 6 | 725 | 87 | |
GenBank mRNA | AJ009672.1 | - | 5 - 6 | 507 | 87 | |
AK175359.1 | - | 5 - 6 | 496 | 87 | ||
AK176882.1 | - | 5 - 6 | 515 | 87 | ||
BT025522.1 | - | 4 - 5 | 441 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |