Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340622.1 | NP_001328567.1 | 1 - 2 | 855 | 85 | |
NM_117023.4 | NP_192693.1 | 2 - 3 | 380 | 85 | ||
GenBank mRNA | AK175417.1 | - | 2 - 3 | 317 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |