Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117033.3 | NP_192703.1 | 1 | 702 | 76 | |
GenBank mRNA | AY065158.1 | - | 1 | 664 | 76 | |
AY087900.1 | - | 1 | 674 | 76 | ||
AY114577.1 | - | 1 | 617 | 76 | ||
AY123002.1 | - | 1 | 617 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |