Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATSPS4F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036532.3 | NP_001031609.1 | 11 - 12 | 2023 | 73 | |
NM_001340652.1 | NP_001329003.1 | 10 - 11 | 2964 | 73 | ||
NM_001340653.1 | NP_001329002.1 | 9 - 10 | 2252 | 73 | ||
NM_001340654.1 | NP_001329004.1 | 7 - 8 | 1282 | 73 | ||
NM_117080.5 | NP_192750.2 | 11 - 12 | 2873 | 73 | ||
GenBank mRNA | KJ138763.1 | - | 10 - 11 | 1845 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |