Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATSPS4F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036532.3 | NP_001031609.1 | 9 - 10 | 1786 | 66 | |
NM_001340652.1 | NP_001329003.1 | 8 - 9 | 2727 | 66 | ||
NM_001340653.1 | NP_001329002.1 | 7 - 8 | 2015 | 66 | ||
NM_001340654.1 | NP_001329004.1 | 5 - 6 | 1045 | 66 | ||
NM_117080.5 | NP_192750.2 | 9 - 10 | 2636 | 66 | ||
GenBank mRNA | KJ138763.1 | - | 8 - 9 | 1608 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |