Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQD16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117132.4 | NP_192802.2 | 2 - 3 | 448 | 71 | |
GenBank mRNA | AK228338.1 | - | 2 - 3 | 443 | 71 | |
BT010145.1 | - | 2 - 3 | 343 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |