Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH9C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340700.1 | NP_001328090.1 | 1 - 2 | 426 | 69 | |
NM_001340701.1 | NP_001328091.1 | 1 - 2 | 415 | 69 | ||
NM_117175.3 | NP_192843.2 | 1 - 2 | 426 | 69 | ||
GenBank mRNA | AY133685.1 | - | 1 - 2 | 201 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |