Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH9C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340700.1 | NP_001328090.1 | 2 - 3 | 640 | 73 | |
NM_001340701.1 | NP_001328091.1 | 2 - 3 | 629 | 73 | ||
NM_117175.3 | NP_192843.2 | 2 - 3 | 640 | 73 | ||
GenBank mRNA | AY133685.1 | - | 2 - 3 | 415 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |