Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117177.3 | NP_192845.1 | 1 - 2 | 559 | 110 | |
NM_179027.1 | NP_849358.1 | 2 - 3 | 258 | 110 | ||
GenBank mRNA | AF442396.1 | - | 1 - 2 | 354 | 110 | |
AY133686.1 | - | 1 - 2 | 354 | 110 | ||
BT001107.1 | - | 2 - 3 | 258 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |