Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOPP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117195.5 | NP_567375.1 | 2 - 3 | 952 | 125 | |
GenBank mRNA | AY086060.1 | - | 2 - 3 | 855 | 125 | |
AY090365.1 | - | 2 - 3 | 899 | 125 | ||
AY122904.1 | - | 2 - 3 | 731 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |