Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340732.1 | NP_001328454.1 | 3 - 4 | 832 | 132 | |
NM_117243.4 | NP_567380.2 | 3 - 4 | 927 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |