Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341318.1 | NP_001329724.1 | 2 - 3 | 484 | 103 | |
NM_118020.5 | NP_193637.2 | 1 - 2 | 663 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |