Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203840.1 | NP_001190769.1 | 4 - 5 | 599 | 84 | |
NM_001341322.1 | NP_001319993.1 | 4 - 5 | 732 | 84 | ||
NM_118022.5 | NP_193639.2 | 4 - 5 | 732 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |