Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118041.5 | NP_193656.2 | 6 - 7 | 701 | 107 | |
GenBank mRNA | AK226939.1 | - | 6 - 7 | 608 | 107 | |
AY099697.1 | - | 6 - 7 | 621 | 107 | ||
BT000298.1 | - | 5 - 6 | 497 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |