Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDG16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118929.5 | NP_194520.3 | 6 - 7 | 2100 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |