Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118962.5 | NP_567801.1 | 8 - 9 | 2500 | 84 | |
GenBank mRNA | BT025339.1 | - | 8 - 9 | 1632 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |