Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118965.4 | NP_567803.1 | 2 - 3 | 645 | 89 | |
GenBank mRNA | AY039986.1 | - | 2 - 3 | 562 | 89 | |
AY133801.1 | - | 2 - 3 | 466 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |