Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118993.4 | NP_194580.1 | 4 - 5 | 593 | 73 | |
GenBank mRNA | AY093228.1 | - | 4 - 5 | 520 | 73 | |
BT000211.1 | - | 3 - 4 | 458 | 73 | ||
U66591.1 | - | 4 - 5 | 528 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |