Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118996.5 | NP_567812.1 | 4 - 5 | 896 | 98 | |
GenBank mRNA | AY099715.1 | - | 4 - 5 | 636 | 98 | |
AY943845.1 | - | 4 - 5 | 347 | 98 | ||
BT000286.1 | - | 4 - 5 | 347 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |