Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HHP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122034.3 | NP_197527.1 | 1 - 2 | 591 | 110 | |
GenBank mRNA | AK226838.1 | - | 1 - 2 | 496 | 110 | |
AY057524.1 | - | 1 - 2 | 503 | 110 | ||
AY086377.1 | - | 1 - 2 | 502 | 110 | ||
AY143975.1 | - | 1 - 2 | 301 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |