Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203419.2 | NP_001190348.1 | 7 - 8 | 1283 | 127 | |
NM_001343652.1 | NP_001330078.1 | 4 - 5 | 451 | 127 | ||
NM_122039.5 | NP_197532.3 | 7 - 8 | 1284 | 127 | ||
GenBank mRNA | DQ118423.1 | - | 7 - 8 | 1284 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |