Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122042.4 | NP_197535.2 | 1 - 2 | 585 | 80 | |
GenBank mRNA | AK228596.1 | - | 1 - 2 | 437 | 80 | |
AY965346.1 | - | 1 - 2 | 254 | 80 | ||
JQ937239.1 | - | 1 - 2 | 254 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |