Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122042.4 | NP_197535.2 | 7 - 8 | 1326 | 96 | |
GenBank mRNA | AK228596.1 | - | 7 - 8 | 1178 | 96 | |
AY965346.1 | - | 7 - 8 | 995 | 96 | ||
JQ937239.1 | - | 7 - 8 | 995 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |