Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161252.2 | NP_001154724.1 | 34 - 35 | 4254 | 103 | |
NM_001343670.1 | NP_001318611.1 | 34 - 35 | 4233 | 103 | ||
NM_001343671.1 | NP_001318612.1 | 35 - 36 | 4686 | 103 | ||
GenBank mRNA | HQ427882.1 | - | 34 - 35 | 4236 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |