Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WAV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122059.4 | NP_568395.1 | 6 - 7 | 888 | 67 | |
GenBank mRNA | AB182157.1 | - | 6 - 7 | 808 | 67 | |
AY080751.1 | - | 6 - 7 | 823 | 67 | ||
AY087569.1 | - | 6 - 7 | 819 | 67 | ||
AY114009.1 | - | 6 - 7 | 689 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |