Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRXL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122061.4 | NP_197554.2 | 2 - 3 | 700 | 66 | |
GenBank mRNA | AK117133.1 | - | 2 - 3 | 646 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Arabidopsis thaliana Probe spans exons |