Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122101.2 | NP_568405.1 | 7 - 8 | 1207 | 93 | |
GenBank mRNA | L23985.1 | - | 7 - 8 | 1167 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |