Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343697.1 | NP_001318615.1 | 2 | 1625 | 90 | |
NM_001343698.1 | NP_001332194.1 | 1 | 1574 | 90 | ||
NM_122120.2 | NP_197611.1 | 1 | 1542 | 90 | ||
GenBank mRNA | AF004214.1 | - | 1 | 1542 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |