Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122138.3 | NP_197625.1 | 1 - 2 | 299 | 79 | |
GenBank mRNA | AY084606.1 | - | 1 - 2 | 213 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |