Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122169.3 | NP_197655.1 | 1 | 1631 | 104 | |
GenBank mRNA | AY058097.1 | - | 1 | 1209 | 104 | |
AY090235.1 | - | 1 | 1208 | 104 | ||
AY149958.1 | - | 1 | 1072 | 104 | ||
DQ411469.1 | - | 1 | 1083 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |