Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CaS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343788.1 | NP_001331124.1 | 4 - 5 | 914 | 91 | |
NM_122212.5 | NP_197697.1 | 4 - 5 | 914 | 91 | ||
GenBank mRNA | AY045798.1 | - | 4 - 5 | 889 | 91 | |
AY079331.1 | - | 4 - 5 | 791 | 91 | ||
AY341888.1 | - | 4 - 5 | 876 | 91 | ||
BT000666.1 | - | 4 - 5 | 893 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |