Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036849.2 | NP_001031926.1 | 1 - 2 | 177 | 65 | |
NM_122214.4 | NP_197699.2 | 1 - 2 | 264 | 65 | ||
GenBank mRNA | AK118129.1 | - | 1 - 2 | 172 | 65 | |
AY518689.1 | - | 1 - 2 | 133 | 65 | ||
BT008588.1 | - | 1 - 2 | 133 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Arabidopsis thaliana Probe spans exons |