Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036849.2 | NP_001031926.1 | 6 - 7 | 603 | 105 | |
NM_122214.4 | NP_197699.2 | 6 - 7 | 690 | 105 | ||
GenBank mRNA | AK118129.1 | - | 6 - 7 | 598 | 105 | |
AY518689.1 | - | 6 - 7 | 559 | 105 | ||
BT008588.1 | - | 6 - 7 | 559 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |