Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343816.1 | NP_001331492.1 | 3 - 4 | 2486 | 77 | |
NM_001343817.1 | NP_001331491.1 | 4 - 5 | 2384 | 77 | ||
NM_001343818.1 | NP_001331490.1 | 4 - 5 | 1942 | 77 | ||
NM_122263.3 | NP_197747.1 | 5 - 6 | 1978 | 77 | ||
GenBank mRNA | BT002944.1 | - | 5 - 6 | 1762 | 77 | |
BT004380.1 | - | 4 - 5 | 1671 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |