Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343827.1 | NP_001330892.1 | 4 - 5 | 1434 | 72 | |
NM_001343828.1 | NP_001330893.1 | 5 - 6 | 1336 | 72 | ||
NM_001343829.1 | NP_001330894.1 | 5 - 6 | 1312 | 72 | ||
NM_122286.4 | NP_197770.1 | 6 - 7 | 1214 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |