Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343834.1 | NP_001330174.1 | 5 - 6 | 1038 | 112 | |
NM_001343835.1 | NP_001330173.1 | 2 - 3 | 681 | 112 | ||
NM_122304.1 | NP_197787.1 | 5 - 6 | 860 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |