Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343836.1 | NP_001332637.1 | 2 - 3 | 358 | 99 | |
NM_122308.5 | NP_197791.1 | 2 - 3 | 488 | 99 | ||
GenBank mRNA | BT015364.1 | - | 2 - 3 | 320 | 99 | |
BT015721.1 | - | 1 - 2 | 178 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |