Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SQP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122319.4 | NP_197802.2 | 3 - 4 | 581 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |