Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SQE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343845.1 | NP_001330191.1 | 4 - 5 | 726 | 90 | |
NM_001343846.1 | NP_001330190.1 | 3 - 4 | 632 | 90 | ||
NM_122321.5 | NP_197804.1 | 4 - 5 | 768 | 90 | ||
GenBank mRNA | AJ005927.1 | - | 4 - 5 | 699 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |