Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SQE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343846.1 | NP_001330190.1 | 5 - 6 | 1127 | 124 | |
NM_122321.5 | NP_197804.1 | 6 - 7 | 1263 | 124 | ||
GenBank mRNA | AJ005927.1 | - | 6 - 7 | 1194 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |