Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203448.2 | NP_001190377.1 | 7 - 8 | 842 | 138 | |
NM_001343861.1 | NP_001331304.1 | 6 - 7 | 3672 | 138 | ||
NM_001343862.1 | NP_001331303.1 | 6 - 7 | 3659 | 138 | ||
NM_001343863.1 | NP_001331302.1 | 5 - 6 | 1125 | 138 | ||
NM_122333.6 | NP_197815.1 | 7 - 8 | 910 | 138 | ||
GenBank mRNA | AF192886.1 | - | 6 - 7 | 481 | 138 | |
AY063993.1 | - | 7 - 8 | 571 | 138 | ||
AY096693.1 | - | 6 - 7 | 481 | 138 | ||
Y18870.1 | - | 594 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |