Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203449.1 | NP_001190378.1 | 3 - 4 | 907 | 124 | |
NM_122336.5 | NP_197818.1 | 3 - 4 | 1131 | 124 | ||
GenBank mRNA | AK226881.1 | - | 3 - 4 | 840 | 124 | |
AY128273.1 | - | 3 - 4 | 829 | 124 | ||
KJ138668.1 | - | 3 - 4 | 763 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |