Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122361.4 | NP_197841.1 | 1 - 2 | 402 | 84 | |
GenBank mRNA | AF386975.1 | - | 1 - 2 | 268 | 84 | |
AY081455.1 | - | 1 - 2 | 197 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |