Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036860.1 | NP_001031937.1 | 1 - 2 | 791 | 103 | |
NM_122386.4 | NP_568454.1 | 1 - 2 | 791 | 103 | ||
GenBank mRNA | AY048282.1 | - | 1 - 2 | 447 | 103 | |
AY050819.1 | - | 1 - 2 | 447 | 103 | ||
AY092991.1 | - | 1 - 2 | 425 | 103 | ||
AY114083.1 | - | 1 - 2 | 411 | 103 | ||
AY114606.1 | - | 1 - 2 | 411 | 103 | ||
BT000761.1 | - | 1 - 2 | 451 | 103 | ||
BT000762.1 | - | 1 - 2 | 449 | 103 | ||
BT006341.1 | - | 1 - 2 | 411 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |